Merck
  • Activation of the apoptotic pathway during prolonged prometaphase blocks daughter cell proliferation.
Activation of the apoptotic pathway during prolonged prometaphase blocks daughter cell proliferation.
Molecular biology of the cell (2018-08-23)
Yumi Uetake, Greenfield Sluder
ABSTRACT
When untransformed human cells spend >1.5 h in prometaphase under standard culture conditions, all daughters arrest in G1 despite normal division of their mothers. We investigate what happens during prolonged prometaphase that leads to daughter cell arrest in the absence of DNA damage. We find that progressive loss of anti-apoptotic MCL-1 activity and oxidative stress act in concert to partially activate the apoptosis pathway, resulting in the delayed death of some daughters and senescence for the rest. At physiological oxygen levels, longer prometaphase durations are needed for all daughters to arrest. Partial activation of apoptosis during prolonged prometaphase leads to persistent caspase activity, which activates the kinase cascade mediating the post-mitotic activation of p38. This in turn activates p53, and the consequent expression of p21stops the cell cycle. This mechanism can prevent cells suffering intractable mitotic defects, which modestly prolong mitosis but allow its completion without DNA damage, from producing future cell generations that are susceptible to the evolution of a transformed phenotype.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Caspase Inhibitor I, InSolution