Merck
  • Chronic administration of diethylnitrosamine to induce hepatocarcinogenesis and to evaluate its synergistic effect with other hepatotoxins in mice.
Chronic administration of diethylnitrosamine to induce hepatocarcinogenesis and to evaluate its synergistic effect with other hepatotoxins in mice.
Toxicology and applied pharmacology (2019-06-10)
Sergio Fuentes-Hernández, Brisa Rodope Alarcón-Sánchez, Dafne Guerrero-Escalera, Alexia Viridiana Montes-Aparicio, María Paulette Castro-Gil, Osiris Germán Idelfonso-García, Sandra Rosas-Madrigal, Diana Ivette Aparicio-Bautista, José Luis Pérez-Hernández, Karina Reyes-Gordillo, M Raj Lakshman, Verónica Rocío Vásquez-Garzón, Rafael Baltiérrez-Hoyos, Ma de Lourdes López-González, Adolfo Sierra-Santoyo, Saúl Villa-Treviño, Julio Isael Pérez-Carreón, Jaime Arellanes-Robledo
ABSTRACT
Hepatocellular carcinoma (HCC) arises after a long period of exposition to etiological factors that might be either independent or collectively contributing. Several rodent models resemble human HCC; however, the major limitation of these models is the lack of chronic injury that reproducibly mimics the molecular alterations as it occurs in humans. Thus, we hypothesized that chronic administration of different DEN treatments identifies the best-fit dose to induce the HCC and/or to determine whether small DEN doses act synergistically with other known hepatotoxins to induce HCC in mice. C57BL/6 J male mice were intraperitoneally injected twice a week for 6 weeks with different DEN doses ranging from 2.5 to 40 mg/kg body weight; then, selected doses (2.5, 5 and 20 mg/kg) for 6, 10, 14, and 18 weeks. We demonstrated that DEN at 20 mg/kg promoted reactive oxygen species and 4-hydroxynonenal production, cell proliferation inflammatory infiltrate, and fibrosis, which in turn induced liver cancer by week 18. These parameters were established by evaluating histopathological changes, HCC markers such as glutathione S-transferase placental-1 (Gstp1), Cytokeratin-19 (Ck19) and prostaglandin reductase-1 (Ptgr1); that of Cyp2e1, a DEN metabolizing enzyme; and the expression of the proliferation marker Ki67. While DEN at 2.5 and 5 mg/kg increased Gstp1 and Ck19, DEN at 20 mg/kg decreased them and Cyp2e1 expression and activity. In summary, our results demonstrate that DEN chronically administrated at 20 mg/kg induces the HCC, while DEN at 2.5 and 5 mg/kg could be useful in elucidating its synergistic effect with other hepatotoxic agents in mice.