Merck
  • Photoluminescent organisms: how to make fungi glow through biointegration with lanthanide metal-organic frameworks.
Photoluminescent organisms: how to make fungi glow through biointegration with lanthanide metal-organic frameworks.
Scientific reports (2019-05-16)
Jeferson Rosário, Leonis L da Luz, Regina Geris, Jéssica G S Ramalho, Antônio F da Silva, Severino Alves Júnior, Marcos Malta
ABSTRACT
We show that filamentous fungi can emit green or red light after the accumulation of particulate lanthanide metal-organic frameworks over the cell wall. These new biohybrids present photoluminescence properties that are unaffected by the components of the cell wall. In addition, the fungal cells internalise lanthanide metal-organic framework particles, storing them into organelles, thereby making these materials promising for applications in living imaging studies.