Merck
  • ELABELA, as a potential diagnostic biomarker of preeclampsia, regulates abnormally shallow placentation via APJ.
ELABELA, as a potential diagnostic biomarker of preeclampsia, regulates abnormally shallow placentation via APJ.
American journal of physiology. Endocrinology and metabolism (2019-03-13)
Lu Zhou, Hang Sun, Ran Cheng, Xiujun Fan, Shanshan Lai, Cheng Deng
ABSTRACT
Preeclampsia (PE) is a major cause of maternal mortality and morbidity worldwide. Although there has been great progress in the understanding of PE, the exact cause for the disease development is still unclear. Recently, studies showed that genetic deletion of ELABELA (ELA, also known as APELA) could induce PE-like symptoms in mice. However, the role of ELA in the disease development of PE remains elusive. Our objective was to measure the changes of ELA levels in maternal serum, urine, and placenta from preeclamptic pregnant women and healthy pregnant women and evaluate the correlation between ELA levels and the occurrence of PE. Additionally, we investigated the effect of ELA on the migration and proliferation of human trophoblast cells. ELA levels are significantly decreased in late-onset PE pregnancies compared with normal pregnancies. The mRNA and protein expressions of ELA and the apelin receptor (APLNR or APJ) in late-onset PE placental tissues are also decreased. Furthermore, our in vitro study showed that the addition of ELA significantly increased the invasion ability and proliferation of trophoblast cells, which were inhibited by the APJ-specific antagonist ML221. Our study identified ELA as significantly decreased in late-onset PE; therefore, it might play an important role in the pathogenesis of late-onset PE.