Merck
  • Matrix metalloproteinase expression in teeth with apical periodontitis is differentially modulated by the modality of root canal treatment.
Matrix metalloproteinase expression in teeth with apical periodontitis is differentially modulated by the modality of root canal treatment.
Journal of endodontics (2010-02-02)
Francisco Wanderley Garcia Paula-Silva, Léa Assed Bezerra da Silva, Yvonne Lorraine Kapila
ABSTRACT
The objective of this study was to investigate the expression of matrix metalloproteinases (MMPs) in apical periodontitis and during the periapical healing phase after root canal treatment. Apical periodontitis was induced in dog teeth, and root canal treatment was performed in a single visit or by using an additional calcium hydroxide root canal dressing. One hundred eighty days after treatment the presence of inflammation was examined, and tissues were stained to detect bacteria. Bacterial status was correlated to the degree of tissue organization, and to further investigate molecules involved in this process, tissues were stained for MMP-1, MMP-2, MMP-8, and MMP-9. Data were analyzed by using one-way analysis of variance followed by Tukey test or Kruskal-Wallis followed by Dunn test. Teeth with apical periodontitis that had root canal therapy performed in a single visit presented an intense inflammatory cell infiltrate. Periapical tissue was extremely disorganized, and this was correlated with the presence of bacteria. Higher MMP expression was evident, similar to teeth with untreated apical periodontitis. In contrast, teeth with apical periodontitis submitted to root canal treatment with calcium hydroxide presented a lower inflammatory cell infiltrate. This group had moderately organized connective tissue, lower prevalence of bacteria, and lower number of MMP-positive cells, similar to healthy teeth submitted to treatment. Teeth treated with calcium hydroxide root canal dressing exhibited a lower percentage of bacterial contamination, a lower MMP expression, and a more organized extracellular matrix, unlike those treated in a single visit. This suggests that calcium hydroxide might be beneficial in tissue repair processes.
Sigma-Aldrich
Anti-MMP-1 (Ab-1) Mouse mAb (41-1E5), liquid, clone 41-1E5, Calbiochem®