Merck
  • Automated enumeration of lymphoid and plasma cells in bone marrow to establish normal reference ranges.
Automated enumeration of lymphoid and plasma cells in bone marrow to establish normal reference ranges.
Journal of clinical pathology (2018-06-03)
James Liang, Jacques A J Malherbe, Kathryn A Fuller, Bob Mirzai, Carly George, Tina L Carter, Catherine H Cole, Belinda B Guo, Katie Meehan, Wendy N Erber
ABSTRACT
The number of precursor and mature lymphoid cells and plasma cells in normal bone marrow trephine (BMT) biopsies throughout the human lifespan is unknown. Reference ranges have been established from aspirated marrow, but due to haemodilution errors, these do not accurately reflect the native marrow milieu. We aimed to define age-specific, normal reference ranges for lymphoid and plasma cells in BMT biopsy specimens using a combined immunophenotyping and digital enumeration approach. Morphologically normal BMT biopsy specimens (n=483) were obtained from patients aged 1 month to 90 years of age. Immunohistochemistry was performed to identify lymphoid progenitors , T-lymphocytes (CD3), B-lymphocytes (CD20) and plasma cells (CD138 and MUM1). Positive cells were counted using digital enumeration software, and the percent positivity for each antigen was determined per case. Mean values were generated for specific age groups, and age-defined reference ranges were determined for each antigen using normalised data. A mean of 16 609 cells (range: 7210-34 097) were counted per biopsy. Infant marrows showed a predominance of immature lymphoid progenitors and B cells. With increasing age, an increase in mean T cell and plasma cell numbers were observed. The results showed the same trends to flow cytometry references for aspirate material although the absolute values differed. Combined immunohistochemistry and automated enumeration gives an accurate, reproducible number of antigen-positive cells and has generated normal reference ranges for these cell types in BMT biopsies. The method and ranges we have established have the potential to be applied in routine clinical practice.
Sigma-Aldrich
1,3-Diiodo-5,5-dimethylhydantoin, ≥96%