Merck
  • Loss of USP28-mediated BRAF degradation drives resistance to RAF cancer therapies.
Loss of USP28-mediated BRAF degradation drives resistance to RAF cancer therapies.
The Journal of experimental medicine (2018-06-09)
Azad Saei, Marta Palafox, Touati Benoukraf, Nishi Kumari, Patrick William Jaynes, Prasanna Vasudevan Iyengar, Eva Muñoz-Couselo, Paolo Nuciforo, Javier Cortés, Christopher Nötzel, Nesaretnam Barr Kumarakulasinghe, John Lalith Charles Richard, Zul Fazreen Bin Adam Isa, Brendan Pang, Marta Guzman, Zhou Siqin, Henry Yang, Wai Leong Tam, Violeta Serra, Pieter Johan Adam Eichhorn
ABSTRACT
RAF kinase inhibitors are clinically active in patients with BRAF (V600E) mutant melanoma. However, rarely do tumors regress completely, with the majority of responses being short-lived. This is partially mediated through the loss of negative feedback loops after MAPK inhibition and reactivation of upstream signaling. Here, we demonstrate that the deubiquitinating enzyme USP28 functions through a feedback loop to destabilize RAF family members. Loss of USP28 stabilizes BRAF enhancing downstream MAPK activation and promotes resistance to RAF inhibitor therapy in culture and in vivo models. Importantly, we demonstrate that USP28 is deleted in a proportion of melanoma patients and may act as a biomarker for response to BRAF inhibitor therapy in patients. Furthermore, we identify Rigosertib as a possible therapeutic strategy for USP28-depleted tumors. Our results show that loss of USP28 enhances MAPK activity through the stabilization of RAF family members and is a key factor in BRAF inhibitor resistance.
Sigma-Aldrich
Anti-USP28 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution