Merck
  • Redox-active iron-containing polymers: synthesis and anionic polymerization of a C-ferrocenyl-substituted phosphaalkene.
Redox-active iron-containing polymers: synthesis and anionic polymerization of a C-ferrocenyl-substituted phosphaalkene.
Chemical communications (Cambridge, England) (2007-08-31)
Kevin J T Noonan, Brian O Patrick, Derek P Gates
ABSTRACT
The addition polymerization of a ferrocenyl-substituted P[double bond]C bond leads to new redox-active polymers with functional ferrocene and phosphine moieties.