Merck
  • Blockade by cAMP of native sodium channels of adult rat skeletal muscle fibers.
Blockade by cAMP of native sodium channels of adult rat skeletal muscle fibers.
The American journal of physiology (1998-12-09)
J F Desaphy, A De Luca, D C Camerino
ABSTRACT
Although the skeletal muscle sodium channel is a good substrate for cAMP-dependent protein kinase (PKA), no functional consequence was observed for this channel expressed in heterologous systems. Therefore, we investigated the effect of 8-(4-chlorophenylthio)adenosine 3',5'-cyclic monophosphate (CPT-cAMP), a membrane-permeable cAMP analog, on the native sodium channels of freshly dissociated rat skeletal muscle fibers by means of the cell-attached patch-clamp technique. Externally applied CPT-cAMP (0.5 mM) reduced peak ensemble average currents by approximately 75% with no change in kinetics. Single-channel conductance and normalized activation curves were unchanged by CPT-cAMP. In contrast, steady-state inactivation curves showed a reduction of the maximal available current and a negative shift of the half-inactivation potential. Similar effects were observed with dibutyryl adenosine 3',5'-cyclic monophosphate but not with cAMP, which does not easily permeate the cell membrane. Incubation of fibers for 1 h with 10 microM H-89, a PKA inhibitor, did not prevent the effect of CPT-cAMP. Finally, the beta-adrenoreceptor agonist isoproterenol mimicked CPT-cAMP when applied at 0.5 mM but had no effect at 0.1 mM. These results indicate that cAMP inhibits native skeletal muscle sodium channels by acting within the fiber, independently of PKA activation.
Sigma-Aldrich
8-(4-Chlorophenylthio)adenosine 3′,5′-cyclic monophosphate sodium salt, ≥97.0% (HPLC), powder