Merck
  • Upregulation of FLG, LOR, and IVL Expression by Rhodiola crenulata Root Extract via Aryl Hydrocarbon Receptor: Differential Involvement of OVOL1.
Upregulation of FLG, LOR, and IVL Expression by Rhodiola crenulata Root Extract via Aryl Hydrocarbon Receptor: Differential Involvement of OVOL1.
International journal of molecular sciences (2018-06-06)
Akiko Hashimoto-Hachiya, Gaku Tsuji, Mika Murai, Xianghong Yan, Masutaka Furue
ABSTRACT
Rhodiola species are antioxidative, salubrious plants that are known to inhibit oxidative stress induced by ultraviolet and γ-radiation in epidermal keratinocytes. As certain phytochemicals activate aryl hydrocarbon receptors (AHR) or OVO-like 1 (OVOL1) to upregulate the expression of epidermal barrier proteins such as filaggrin (FLG), loricrin (LOR), and involucrin (IVL), we investigated such regulation by Rhodiola crenulata root extract (RCE). We demonstrated that RCE induced FLG and LOR upregulation in an AHR-OVOL1-dependent fashion. However, RCE-mediated IVL upregulation was AHR-dependent but OVOL1-independent. Coordinated upregulation of skin barrier proteins by RCE via AHR may be beneficial in the management of barrier-disrupted inflammatory skin diseases such as atopic dermatitis.
Sigma-Aldrich
MISSION® esiRNA, targeting human AHR, RP11-507K12.1
Sigma-Aldrich
MISSION® esiRNA, targeting human OVOL1