Merck
Calcium homeostasis: solving the solubility problem.
Journal of musculoskeletal & neuronal interactions (2006-12-23)
R V Talmage, D W Talmage
ABSTRACT
This report summarizes the evidence that the control of the concentration of free calcium ions in body fluids is centered at mineralized bone surfaces. This process involves an increase in the solubility of bone mineral produced by the non-collagenous proteins existing in the bone extracellular fluid (ECF) and on the adjacent surfaces of bone. The result is a basic equilibrium level produced in the absence of parathyroid hormone (PTH), which is well above the solubility of bone mineral. The effect of PTH is to increase the solubility of bone mineral still further, but the mechanism by which the hormone acts is unknown. The lining cells of the bone contain receptors for PTH and can be observed to respond to this hormone, but the relationship between this response and the increased solubility of bone remains to be discovered. Further research in this field is strongly urged.
Sigma-Aldrich
SPARC/Osteonectin human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture