Merck
  • Long Electron-Hole Diffusion Length in High-Quality Lead-Free Double Perovskite Films.
Long Electron-Hole Diffusion Length in High-Quality Lead-Free Double Perovskite Films.
Advanced materials (Deerfield Beach, Fla.) (2018-04-01)
Weihua Ning, Feng Wang, Bo Wu, Jun Lu, Zhibo Yan, Xianjie Liu, Youtian Tao, Jun-Ming Liu, Wei Huang, Mats Fahlman, Lars Hultman, Tze Chien Sum, Feng Gao
ABSTRACT
Developing environmentally friendly perovskites has become important in solving the toxicity issue of lead-based perovskite solar cells. Here, the first double perovskite (Cs2 AgBiBr6 ) solar cells using the planar structure are demonstrated. The prepared Cs2 AgBiBr6 films are composed of high-crystal-quality grains with diameters equal to the film thickness, thus minimizing the grain boundary length and the carrier recombination. These high-quality double perovskite films show long electron-hole diffusion lengths greater than 100 nm, enabling the fabrication of planar structure double perovskite solar cells. The resulting solar cells based on planar TiO2 exhibit an average power conversion efficiency over 1%. This work represents an important step forward toward the realization of environmentally friendly solar cells and also has important implications for the applications of double perovskites in other optoelectronic devices.
Sigma-Aldrich
Titanium(IV) isopropoxide, 99.999% trace metals basis