Merck
  • Endocycle-related tubular cell hypertrophy and progenitor proliferation recover renal function after acute kidney injury.
Endocycle-related tubular cell hypertrophy and progenitor proliferation recover renal function after acute kidney injury.
Nature communications (2018-04-11)
Elena Lazzeri, Maria Lucia Angelotti, Anna Peired, Carolina Conte, Julian A Marschner, Laura Maggi, Benedetta Mazzinghi, Duccio Lombardi, Maria Elena Melica, Sara Nardi, Elisa Ronconi, Alessandro Sisti, Giulia Antonelli, Francesca Becherucci, Letizia De Chiara, Ricardo Romero Guevara, Alexa Burger, Beat Schaefer, Francesco Annunziato, Hans-Joachim Anders, Laura Lasagni, Paola Romagnani
ABSTRACT
Acute kidney injury (AKI) is considered largely reversible based on the capacity of surviving tubular cells to dedifferentiate and replace lost cells via cell division. Here we show by tracking individual tubular cells in conditional Pax8/Confetti mice that kidney function is  recovered after AKI despite substantial tubular cell loss. Cell cycle and ploidy analysis upon AKI in conditional Pax8/FUCCI2aR mice and human biopsies identify endocycle-mediated hypertrophy of tubular cells. By contrast, a small subset of Pax2+ tubular progenitors enriches via higher stress resistance and clonal expansion and regenerates necrotic tubule segments, a process that can be enhanced by suitable drugs. Thus,  renal functional recovery upon AKI involves remnant tubular cell hypertrophy via endocycle and limited progenitor-driven regeneration that can be pharmacologically enhanced.
Sigma-Aldrich
Anti-Aquaporin 1 Antibody, 1.0 mg/mL, Chemicon®