- Effects of simvastatin and taurine on delayed cerebral vasospasm following subarachnoid hemorrhage in rabbits.
Effects of simvastatin and taurine on delayed cerebral vasospasm following subarachnoid hemorrhage in rabbits.
The aim of the current study was to observe the effects of simvastatin and taurine on delayed cerebral vasospasm (DCVS) following experimental subarachnoid hemorrhage (SAH) in rabbits. A total of 48 New Zealand white rabbits were allocated at random into four groups (control, SAH, SAH + simvastatin and SAH + taurine groups; n=12 each). The rabbit model of DCVS was established using a double hemorrhage method, which involved injecting autologous arterial blood into the cisterna magna in the SAH groups. The SAH + simvastatin group was administered oral simvastatin (5 mg/kg) daily between days 0-6. The SAH + taurine group was administered oral taurine (50 mg/kg) daily between days 0-6. Starch (50 mg/kg) was administered orally to the animals in the other two groups (control and SAH groups). The control group were not subjected to any other injections or treatment. The internal diameter and internal diameter/wall thickness of the basilar artery (BA) were measured. The expression levels of tumor necrosis factor (TNF)-α, interleukin (IL)-1β and IL-6 were determined using immunohistochemical and quantitative polymerase chain reaction methods following the sacrifice of all animals on day 7. The activity of nuclear factor (NF)-κB in the BA was also measured using an electrophoretic mobility shift assay. The BA walls in the SAH + simvastatin and SAH + taurine groups exhibited reduced narrowing and corrugation of the tunica elastica interna compared with the SAH group. At the protein and cDNA levels, it was found that cerebral vasospasm of the BA in the SAH + simvastatin and SAH + taurine groups was alleviated, as indicated by the reduced expression of TNF-α, IL-1β, IL-6 and NF-κB compared with the SAH group (P<0.05). In conclusion, simvastatin and taurine reduced DCVS following SAH in rabbits, which suggests that these compounds may exert anti-inflammatory effects.