![TSKgel® SuperSW Guard Column phase diol, L × I.D. 3.5 cm × 4.6 mm, 4 μm particle size](/deepweb/assets/sigmaaldrich/product/images/409/236/c7921794-6b2a-465f-8f54-0eb26b9d49b5/640/c7921794-6b2a-465f-8f54-0eb26b9d49b5.jpg)
TSKgel® SuperSW (4 µm) HPLC Column
![TSKgel® SuperSW Guard Column phase diol, L × I.D. 3.5 cm × 4.6 mm, 4 μm particle size](/deepweb/assets/sigmaaldrich/product/images/409/236/c7921794-6b2a-465f-8f54-0eb26b9d49b5/640/c7921794-6b2a-465f-8f54-0eb26b9d49b5.jpg)
Synonym(s):
TSKgel® Size Exclusion (SW-Type) HPLC Guard Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing