![Tri-n-butyl phosphate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/384/284/e813e116-3b16-44f3-b0a0-712de492bcb5/640/e813e116-3b16-44f3-b0a0-712de492bcb5.png)
Tri-n-butyl phosphate
![Tri-n-butyl phosphate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/384/284/e813e116-3b16-44f3-b0a0-712de492bcb5/640/e813e116-3b16-44f3-b0a0-712de492bcb5.png)
Synonym(s):
Tributyl phosphate, TBP, TBPA
Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing