Skip to Content
Merck
trans-2-Hexenyl hexanoate ≥93%, natural, FG

trans-2-Hexenyl hexanoate

trans-2-Hexenyl hexanoate ≥93%, natural, FG
Synonym(s):
2-Hexenyl hexanoate
Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing