![Topiramate impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/269/460/c9eaa448-63c2-4eea-b568-080e4d030d1b/640/c9eaa448-63c2-4eea-b568-080e4d030d1b.png)
Topiramate impurity A
![Topiramate impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/269/460/c9eaa448-63c2-4eea-b568-080e4d030d1b/640/c9eaa448-63c2-4eea-b568-080e4d030d1b.png)
Synonym(s):
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose, D-Fructopyranose diacetonide
Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
Products
Formula Weight
Manufacturer
Quality Segment
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing