Skip to Content
NL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 302
Products
Manufacturer
Physical Form
Diameter (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF84288550
Stainless steel - AISI 302,
foil, Fe/Cr18%/Ni 8%, thickness 0.025 mm, diameter 15 mm, temper hard
Expand
View Pricing
GF83109236
Stainless steel - AISI 302,
foil, thickness 0.025 mm, Fe/Cr18%/Ni 8%, size 100 × 100 mm, temper hard
Expand
View Pricing
GF26274304
Stainless steel - AISI 302,
foil, Fe/Cr18%/Ni 8%, thickness 0.025 mm, 10 mm diameter, temper hard
Expand
View Pricing
GF30033379
Stainless steel - AISI 302,
foil, Fe/Cr18%/Ni 8%, thickness 0.025 mm, 4 mm diameter, temper hard
Expand
View Pricing
GF54171288
Stainless steel - AISI 302,
foil, Fe/Cr18%/Ni 8%, thickness 0.025 mm, 6 mm diameter, temper hard
Expand
View Pricing
GF36068913
Stainless steel - AISI 302,
foil, Fe/Cr18%/Ni 8%, thickness 0.025 mm, 25 mm diameter, temper hard
Expand
View Pricing
GF08614009
Stainless steel - AISI 302,
foil, Fe/Cr18%/Ni 8%, thickness 0.025 mm, 50 mm diameter, temper hard
Expand
View Pricing
GF71410193
Stainless steel - AISI 302,
foil, thickness 0.025 mm, Fe/Cr18%/Ni 8%, diameter 8 mm, temper hard
Expand
View Pricing
GF88654720
Stainless steel - AISI 302,
foil, thickness 0.025 mm, Fe/Cr18%/Ni 8%, size 150 × 150 mm, temper hard
Expand
View Pricing