Salicortin
Empirical Formula (Hill Notation):
C20H24O10
CAS Number:
Molecular Weight:
424.40
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing