
Rottlerin

Synonym(s):
Rottlerin
Empirical Formula (Hill Notation):
C30H28O8
CAS Number:
Molecular Weight:
516.54
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing