![Primulaverin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/551/890/6955c479-ae29-420c-9073-c93db27c3faf/640/6955c479-ae29-420c-9073-c93db27c3faf.png)
Primulaverin
![Primulaverin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/551/890/6955c479-ae29-420c-9073-c93db27c3faf/640/6955c479-ae29-420c-9073-c93db27c3faf.png)
Synonym(s):
5-Methoxy-2-[(6-O-β-D-xylopyranosyl-β-D-glucopyranosyl)oxy]benzoic acid methyl ester
Empirical Formula (Hill Notation):
C20H28O13
CAS Number:
Molecular Weight:
476.43
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing