![Pralidoxime Chloride certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/163/436/a3640822-f172-431c-a174-cf2141a24109/640/a3640822-f172-431c-a174-cf2141a24109.jpg)
Pralidoxime Chloride
![Pralidoxime Chloride certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/163/436/a3640822-f172-431c-a174-cf2141a24109/640/a3640822-f172-431c-a174-cf2141a24109.jpg)
Synonym(s):
(NE)-N-[(1-methylpyridin-1-ium-2-yl)methylidene]hydroxylamine;chloride
Empirical Formula (Hill Notation):
C7H9ClN2O
CAS Number:
Molecular Weight:
172.61
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing