Skip to Content
Merck
vibrant-m

Perindopril Related Compound D

vibrant-m
Synonym(s):
S,3S,5aS,9aS,10aR)-Decahydro-3-methyl-1,4-dioxo-α-propylpyrazino[1,2-a]indole-2(1H)-acetic acid, (S)-2-{(3S,5aS,9aS,10aR)-3-Methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl}pentanoic acid, Perindoprilat lactam B
Empirical Formula (Hill Notation):
C17H26N2O4
CAS Number:
Molecular Weight:
322.40

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1