perfluorooctanesulfonamide
Synonym(s):
1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-1-octanesulfonamide, Perfluorooctylsulfonamide
Empirical Formula (Hill Notation):
C8H2F17NO2S
CAS Number:
Molecular Weight:
499.14
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2