Skip to Content
Merck
Octanal analytical standard

Octanal

Octanal analytical standard
Synonym(s):
Aldehyde C8, Caprylic aldehyde, Octyl aldehyde, Octanal
Linear Formula:
CH3(CH2)6CHO
CAS Number:
Molecular Weight:
128.21
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing