Skip to Content
Merck
N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine N-oxide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine N-oxide

N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine N-oxide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C15H19N5O
CAS Number:
Molecular Weight:
285.34

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade