Skip to Content
Merck
Netilmicin sulfate salt

Netilmicin sulfate salt

Netilmicin sulfate salt
Synonym(s):
1-N-Ethylsisomicin
Linear Formula:
C21H41N5O7 · 5/2H2SO4
CAS Number:
Molecular Weight:
720.78
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing