Skip to Content
Merck
vibrant-m

Methyl N-4-[2-(5-chloro-2-methoxybenzamido)ethyl]benzenesulfonylcarbamate

vibrant-m
Synonym(s):
Methyl {{4-{2-[(5-chloro-2-methoxybenzoyl)amino]ethyl}phenyl}sulfonyl}carbamate
Empirical Formula (Hill Notation):
C18H19ClN2O6S
CAS Number:
Molecular Weight:
426.87

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade