Skip to Content
Merck
Methyl 3-aminobenzoate ≥97.0% (GC)

Methyl 3-aminobenzoate

Methyl 3-aminobenzoate ≥97.0% (GC)
Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing