Skip to Content
Merck
Methyl 2-pyrrolyl ketone ≥98%, FG

Methyl 2-pyrrolyl ketone

Methyl 2-pyrrolyl ketone ≥98%, FG
Synonym(s):
2-Acetylpyrrole, Methyl 2-pyrrolyl ketone
Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing