![Medronic acid impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/832/1460f52c-2582-46ac-a330-239db7a0ed84/640/1460f52c-2582-46ac-a330-239db7a0ed84.png)
Medronic acid impurity B
![Medronic acid impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/832/1460f52c-2582-46ac-a330-239db7a0ed84/640/1460f52c-2582-46ac-a330-239db7a0ed84.png)
Synonym(s):
Tetraisopropyl methylenediphosphonate
Linear Formula:
CH2[P(O)[OCH(CH3)2]2]2
CAS Number:
Molecular Weight:
344.32
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing