
Losartan Related Compound C

Synonym(s):
2-Butyl-4-chloro-1-[[2′-(1H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxaldehyde, 2-Butyl-4-chloro-1-[[2′-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazole-5-carbaldehyde, EXP-3179
Empirical Formula (Hill Notation):
C22H21ClN6O
CAS Number:
Molecular Weight:
420.89
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2