Kojibiose
Synonym(s):
α-D-Glc-(1→2)-D-Glc, 2-O-α-D-Glucopyranosyl-D-glucose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing