Skip to Content
Merck
vibrant-m

Indinavir for system suitability

vibrant-m
Synonym(s):
Indinavir monohydrate
Empirical Formula (Hill Notation):
C36H47N5O4 · H2O
CAS Number:
Molecular Weight:
631.80

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1