Skip to Content
Merck
HS161 Umicore

HS161

HS161 Umicore
Synonym(s):
Trimethyl(methylcyclopentadienyl)platinum(IV)
Linear Formula:
C5H4CH3Pt(CH3)3
CAS Number:
Molecular Weight:
319.30

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing