Skip to Content
Merck
Glyceryl 1,3-dioctadecanoate-2-octanoate-1-13C endotoxin tested, ≥99 atom % 13C, ≥99% (CP)

Glyceryl 1,3-dioctadecanoate-2-octanoate-1-13C

Glyceryl 1,3-dioctadecanoate-2-octanoate-1-13C endotoxin tested, ≥99 atom % 13C, ≥99% (CP)
Synonym(s):
13C Labeled glyceryl 1,3-dioctadecanoate-2-octanoate, Glyceryl 1,3-distearate-3-ocatanoate-1-13C
Empirical Formula (Hill Notation):
13CC46H90O6
Molecular Weight:
752.21

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group