Skip to Content
Merck
Glucagon ≥95% (HPLC), powder, synthetic

Glucagon

Glucagon ≥95% (HPLC), powder, synthetic
Empirical Formula (Hill Notation):
C153H225N43O49S
CAS Number:
Molecular Weight:
3482.75

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing