![Eosin Y solution 5 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/380/722/a9122f48-f75f-44f9-b393-8709bb6484d6/640/a9122f48-f75f-44f9-b393-8709bb6484d6.png)
Eosin Y solution
![Eosin Y solution 5 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/380/722/a9122f48-f75f-44f9-b393-8709bb6484d6/640/a9122f48-f75f-44f9-b393-8709bb6484d6.png)
Synonym(s):
2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid red 87, Eosin yellowish solution
Empirical Formula (Hill Notation):
C20H6Br4Na2O5
CAS Number:
Molecular Weight:
691.85
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing