Skip to Content
Merck
Diacetylmorphine Hydrochloride certified reference material, pharmaceutical secondary standard

Diacetylmorphine Hydrochloride

Diacetylmorphine Hydrochloride certified reference material, pharmaceutical secondary standard
Synonym(s):
Heroin hydrochloride
Empirical Formula (Hill Notation):
C21H23NO5 · HCl
CAS Number:
Molecular Weight:
405.87

Products

Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1