Skip to Content
Merck
Compressible sugar United States Pharmacopeia (USP) Reference Standard

Compressible sugar

Compressible sugar United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Sucrose, α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing