![Chlorambucil British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/352/952/8a183cb8-7541-4fa8-99fa-c805d3b03e7e/640/8a183cb8-7541-4fa8-99fa-c805d3b03e7e.png)
Chlorambucil
![Chlorambucil British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/352/952/8a183cb8-7541-4fa8-99fa-c805d3b03e7e/640/8a183cb8-7541-4fa8-99fa-c805d3b03e7e.png)
Synonym(s):
4-(4-[Bis(2-chloroethyl)amino]phenyl)butyric acid, 4-[Bis(2-chloroethyl)amino]benzenebutyric acid
Empirical Formula (Hill Notation):
C14H19Cl2NO2
CAS Number:
Molecular Weight:
304.21
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing