Skip to Content
Merck
Benzothiazole analytical standard

Benzothiazole

Benzothiazole analytical standard
Empirical Formula (Hill Notation):
C7H5NS
CAS Number:
Molecular Weight:
135.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing