![Anti-UQCRC1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/314/267/1fc5f0a0-9c47-41c2-afa0-4ff4bf966ad1/640/1fc5f0a0-9c47-41c2-afa0-4ff4bf966ad1.jpg)
Anti-UQCRC1 antibody produced in rabbit
![Anti-UQCRC1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/314/267/1fc5f0a0-9c47-41c2-afa0-4ff4bf966ad1/640/1fc5f0a0-9c47-41c2-afa0-4ff4bf966ad1.jpg)
Synonym(s):
Anti-Core I protein antibody produced in rabbit, Anti-Ubiquinol-cytochrome-c reductase complex core protein 1, mitochondrial precursor antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing