Albuterol Related Compound E
Synonym(s):
2,2′-Oxybis(methylene)bis{4-[2-(tert-butylamino)-1-hydroxyethyl]phenol} diacetate diacetate, Sambutamol dimer diacetate
Empirical Formula (Hill Notation):
C26H40N2O5 · 2C2H4O2
Molecular Weight:
580.71
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2