![Adenosine-5′-diphosphoglucose disodium salt ≥93%](/deepweb/assets/sigmaaldrich/product/structures/186/119/26048bf4-68af-4d1a-8972-062aed149b17/640/26048bf4-68af-4d1a-8972-062aed149b17.png)
Adenosine-5′-diphosphoglucose disodium salt
![Adenosine-5′-diphosphoglucose disodium salt ≥93%](/deepweb/assets/sigmaaldrich/product/structures/186/119/26048bf4-68af-4d1a-8972-062aed149b17/640/26048bf4-68af-4d1a-8972-062aed149b17.png)
Synonym(s):
ADP-Glucose, ADP-glc, ADPG
Empirical Formula (Hill Notation):
C16H23N5Na2O15P2
CAS Number:
Molecular Weight:
633.31
Products
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing