![4α-Phorbol 12,13-didecanoate solid](/deepweb/assets/sigmaaldrich/product/structures/396/133/a260f54a-0d11-41ae-9313-dde9ca06d099/640/a260f54a-0d11-41ae-9313-dde9ca06d099.png)
4α-Phorbol 12,13-didecanoate
![4α-Phorbol 12,13-didecanoate solid](/deepweb/assets/sigmaaldrich/product/structures/396/133/a260f54a-0d11-41ae-9313-dde9ca06d099/640/a260f54a-0d11-41ae-9313-dde9ca06d099.png)
Synonym(s):
4α-Phorbol 12-13-dicaprinate, 4αPDD, 4α-Phorbol-12,13-didecanoate
Empirical Formula (Hill Notation):
C40H64O8
CAS Number:
Molecular Weight:
672.93
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing