Skip to Content
Merck
2-Phenylethyl β-D-thiogalactoside ≥98% (TLC)

2-Phenylethyl β-D-thiogalactoside

2-Phenylethyl β-D-thiogalactoside ≥98% (TLC)
Synonym(s):
PETG, Phenethyl β-D-thiogalactoside
Empirical Formula (Hill Notation):
C14H20O5S
CAS Number:
Molecular Weight:
300.37

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing