Product Name
5-Phenyl-1,3,4-oxadiazole-2-propionic acid 97%
Product Number
663409
Product Brand
aldrich
CAS Number
23464-98-4
Molecular Weight
218.21
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN POWDER, CRYSTALS OR CRYSTALLINE |
POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 58.4% - 62.7% |
NITROGEN 12.3% - 13.3% | |
AND/OR | |
TITRATION | 96.5% - 103.5% (WITH NAOH) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED JANUARY 2006 |