Skip to Content
Merck
All Photos(1)

Documents

286702

Sigma-Aldrich

Mevalonic lactone

97%

Synonym(s):

β-Hydroxy-β-methyl-δ-valerolactone, (±)-β-Hydroxy-β-methyl-δ-valerolactone, (±)-3-Hydroxy-3-methyl δ-valerolactone, (±)-Mevalolactone, (±)-Mevalonic acid lactone, (±)-Mevalonolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein:
80960
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

refractive index

n20/D 1.473 (lit.)

bp

145-150 °C/5 mmHg (lit.)

mp

28 °C (lit.)

SMILES string

CC1(O)CCOC(=O)C1

Other Notes

Contains mevalonic acid

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service